Learn More
Serpin F2/alpha 2-Antiplasmin Rat anti-Mouse, Clone: 209126, R&D Systems™
Rat Monoclonal Antibody
Marke:  RD Systems (Bio-Techne) MAB1239
Weitere Details : Netto-Gewicht : 0.00010kg
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 13418485
  • 377.53 € / 500 Mikrogramm
Voraussichtlicher Liefertermin 30-07-2019
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
Serpin F2/alpha 2-Antiplasmin Monoclonal antibody MAB1239 Specifically detects Serpin F2/alpha 2-Antiplasmin in Mouse samples. It is validated for Western Blotting
Spezifikation
Spezifikation
Serpin F2/alpha 2-Antiplasmin
209126
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
SERPINF2
Mouse myeloma cell line NS0-derived recombinant mouse Serpin F2/alpha 2 -Antiplasmin Val28-Lys491 Accession # Q61247
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
5345
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q61247
AAPclade F (alpha-2 antiplasmin, alpha 2-Antiplasmin, Alpha-2-AP, Alpha-2-PI, Alpha-2-plasmin inhibitor, pigment epithelium derived factor), member 2, Serpin F2, serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epitheliumderived factor), member 2
Rat
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse Serpin F2/alpha 2 -Antiplasmin in Western blots and direct ELISAs. In direct ELISAs, 100% cross-reactivity with recombinant human (rh) Serpin F2 and no cross-reactivity with rhSerpin A1, A3, A4 or recombinant mouse Serpin A5, C1, or D1 is observed.