missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Serum Albumin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen R01011E
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Purity: IgG is > 97.5% total protein by SDS-PAGE. Becker titer results showed > 1.8 mg/mL (antigen/antibody).
Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough `produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.Spezifikation
Human Serum Albumin | |
Polyclonal | |
Unconjugated | |
ALB | |
Alb; Alb1; Alb-1; albumin; albumin (32 AA); albumin (AA 34); albumin 1; Albza; ANALBA; cell growth inhibiting protein 42; DKFZp779N1935; FDAH; GIG20; GIG42; growth-inhibiting protein 20; PRO0883; PRO0903; PRO1341; PRO1708; PRO2044; PRO2619; PRO2675; Serum albumin; UNQ696/PRO1341 | |
Rabbit | |
Protein A | |
RUO | |
213 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunodiffusion, Immunoelectrophoresis, Western Blot | |
1.0 mg/mL | |
PBS with 0.02% sodium azide; pH 7.4 | |
P02768 | |
ALB | |
Purified human serum albumin | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |