missing translation for 'onlineSavingsMsg'
Learn More

SIN3A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA1870

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11543831

  • 485.00 € / 100 Mikrogramm
Verfügbar ab: 16-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA1-870 detects mSin3A from human and mouse tissues and cells. PA1-870 has been successfully used in Western blot, ICC/IF ChIP, IHC-P and immunoprecipitation procedures. By Western blot, this antibody detects an ∽150 kDa protein representing mSin3A from T47D cell extract. PA1-870 immunogen is a synthetic peptide corresponding to residues M(1) K R R L D D Q E S P V Y A A Q Q R R(19) of mouse mSin3A.

SIN3A Polyclonal antibody specifically detects SIN3A in Human, Mouse samples. It is validated for ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

SIN3A
Polyclonal
Unconjugated
SIN3A
AW553200; Histone deacetylase complex subunit Sin3a; Kiaa4126; Mammalian Sin3A; mKIAA4126; mSin3A; Paired amphipathic helix protein Sin3a; SIN; Sin3; SIN3 homolog A, transcription regulator; SIN3 transcription regulator family member A; SIN3 transcription regulator homolog A; SIN3A; transcriptional corepressor Sin3a; transcriptional co-repressor Sin3A; transcriptional regulator, SIN3 yeast homolog A; transcriptional regulator, SIN3A; transcriptional regulator, SIN3A (yeast)
Rabbit
Antigen affinity chromatography
RUO
20466, 25942
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ChIP Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q60520, Q96ST3
SIN3A
Synthetic peptide corresponding to residues M(1) K R R L D D Q E S P V Y A A Q Q R R(19) of mouse mSin3A.
100 μg
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote