missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIRT7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA513226
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine, mouse and rat based on sequence homology.
SIRT7 Polyclonal antibody specifically detects SIRT7 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
SIRT7 | |
Polyclonal | |
Unconjugated | |
SIRT7 | |
MGC126840; MGC126842; NAD-dependent deacetylase sirtuin-7; NAD-dependent protein deacetylase sirtuin-7; NAD-dependent protein deacylase sirtuin-7; regulatory protein SIR2 homolog 7; silent mating type information regulation 2, S.cerevisiae, homolog 7; Sir2l7; SIR2-like protein 7; sir2-related protein type 7; Sirt7; sirtuin 7; sirtuin type 7; Unknown (protein for MGC:142712) | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51547 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q9NRC8 | |
SIRT7 | |
KLH conjugated synthetic peptide between 331-360 amino acids from the C-terminal region of human SIRT7 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |