missing translation for 'onlineSavingsMsg'
Learn More

Smooth Muscle Actin, clone: 1A4, Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA137027

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11545172

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen »


Beschreibung

Beschreibung

This antibody stains smooth muscle cells in vessel walls, gut wall, and myometrium. Myoepithelial cells in breast and salivary gland are also stained. It reacts with tumors arising from smooth muscles and myoepithelial cells. Actin from fibroblasts (beta- and gamma-cytoplasmic), striated muscle (alpha-sarcomeric), and myocardium (alpha-myocardial) are not stained.

This antibody is predicted to react with mouse, rat, rabbit, baboon, and bovine based on sequence homology.A suggested positive control is small intestine (smooth muscle) or leiomyoma.
Spezifikation

Spezifikation

Smooth Muscle Actin
Monoclonal
Unconjugated
P62736
ACTA2
Synthetic peptide from N-terminus of human alpha smooth muscle isoform
1 mL
Primary
Human
Antibody
IgG1 κ
Immunohistochemistry (Paraffin)
1A4
ACTA2
AAT6, ACTSA, alpha-sma
Mouse
Protein A
RUO
59
Store at 4°C, store in dark.
Purified
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote