Learn More
SMRT Mouse anti-Human, Unlabeled, Clone: 44, BD
Mouse Monoclonal Antibody
335.00 € - 629.00 €
Spezifikation
Antigen | SMRT |
---|---|
Klon | 44 |
Konzentration | 250μg/mL |
Anwendungen | Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15875909
|
BD Biosciences
611386 |
50 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
15885909
|
BD Biosciences
611387 |
150 μg |
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt. Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen |
||||||
Beschreibung
Steroid nuclear hormone receptors regulate physiological homeostasis through repression and activation of gene transcription. In the absence of hormone, DNA-bound steroid receptors recruit corepressor, such as silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) and nuclear receptorcorepressor (N-CoR), which bind to the free ligand binding domain of the thyroid hormone receptor, retinoic acid receptor, and other nuclear receptors. SMRT and N-CoR form a large protein complex with histone deacetylase I (HDAC1) and Sin3A, which deacetylates histones to alter chromatin structure in a manner that inhibits transcription. In addition, SMRT and N-CoR proteins interact with the transcription factor CBF/RBP-Jk to regulate Notch signaling pathways. SMRT contains four domains related to repressor activity (RD1, RD2, SRD1, and SRD2), two receptor interaction domains (RID1 and RID2), and a SANT region that is found in yeast chromatin remodeling factor SWI3. The ubiquitous expression pattern and corepressor activity of SMRT indicates that this protein is important for hormonal control of gene transcription in many tissues.
Immunofluorescence, Western Blotting
Spezifikation
SMRT | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
44 | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Human SMRT aa. 1366-1473 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.