missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SSX5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA530180
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SSX5 Polyclonal antibody specifically detects SSX5 in Human samples. It is validated for Western Blot.Spezifikation
SSX5 | |
Polyclonal | |
Unconjugated | |
SSX5 | |
protein SSX5; SSX family member 5; SSX5; synovial sarcoma, X breakpoint 5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6758 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.61 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O60225 | |
SSX5 | |
Recombinant protein fragment corresponding to a region within amino acids 13 and 214 of Human SSX5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |