Learn More
ST18 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA540764
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VIKKGKVSEE LMTIKLKATG GIESDEEIRH LDEEIKELNE SNLKIEADMM Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
ST18 Polyclonal antibody specifically detects ST18 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
ST18 | |
Polyclonal | |
Unconjugated | |
ST18 | |
AV348974; C2-HC type zinc finger protein r-MyT3; Kiaa0535; mKIAA0535; Myt3; Neural zinc finger factor 3; neural zinc finger protein NZF-3; neural zinc finger transcription factor 3; Nzf3; NZF-3; r-MyT3; St18; ST18 C2H2C-type zinc finger transcription factor; ST18, C2H2C-type zinc finger; suppression of tumorigenicity 18; suppression of tumorigenicity 18 (breast carcinoma) (zinc finger protein); suppression of tumorigenicity 18 protein; suppression of tumorigenicity 18, zinc finger; suppression of tumorigenicity protein 18; ZC2H2C3; ZC2HC10; zinc finger protein 387; ZNF387 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9705 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O60284 | |
ST18 | |
Synthetic peptide directed towards the middle region of human ST18 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |