missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST6GALNAC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA531200
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HeLa, HepG2, HCT116. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ST6GALNAC1 Polyclonal antibody specifically detects ST6GALNAC1 in Human samples. It is validated for Western Blot.Spezifikation
ST6GALNAC1 | |
Polyclonal | |
Unconjugated | |
ST6GALNAC1 | |
alpha-2,6-sialyltransferase ST6GalNAc I; alpha-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; GalNAc alpha-2, 6-sialyltransferase I, long form; galNAc alpha-2,6-sialyltransferase I; HSY11339; sialyltransferase 7 ((alpha-N-acetylneuraminyl 2,3-beta-galactosyl-1,3)-N-acetyl galactosaminde alpha-2,6-sialyltransferase) A; sialyltransferase 7 ((alpha-N-acetylneuraminyl-2,3-beta-galactosyl-1,3)-N-acetyl galactosaminide alpha-2,6-sialyltransferase) A; sialyltransferase 7A; SIAT7A; SIAT7-A; ST6 (alpha-N-acetyl-neuraminyl-2,3-beta-galactosyl-1,3)-N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6 GalNAc alpha-2,6-sialyltransferase 1; ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1; ST6GalNAc I; ST6GALNAC1; ST6GALNAC1, alpha-2,6-sialyltransferase 1; ST6GalNAcI; STYI; UNQ543/PRO848 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55808 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NSC7 | |
ST6GALNAC1 | |
Recombinant protein fragment corresponding to a region within amino acids 175 and 416 of Human ST6GALNAC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |