missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Strep Tag Monoclonal Antibody (GT517), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA517282
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: strep-tagged protein expressing 293T cells. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Strep Tag Monoclonal antibody specifically detects Strep Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
Strep Tag | |
Monoclonal | |
1 mg/mL | |
PBS with 20% glycerol and no preservative; pH 7 | |
Immunogen Amino acid sequence - ASWSHPQFEKGA | |
100 μL | |
Primary | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
GT517 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
Tag |