missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUFU Monoclonal Antibody (C.60.7), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA514956
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
It is not recommended to aliquot this antibody.
SUFU Monoclonal antibody specifically detects SUFU in Human, Mouse, Non-human Primate, Rat samples. It is validated for Western Blot.Spezifikation
SUFU | |
Monoclonal | |
Unconjugated | |
SUFU | |
OTTHUMP00000020377; OTTHUMP00000020378; OTTHUMP00000020379; PRO1280; RP11-47A8.1; Su(fu); Sufu; SUFU negative regulator of hedgehog signaling; SUFUH; SUFUXL; suppressor of fused homolog; suppressor of fused homolog (Drosophila); UNQ650/PRO1280 | |
Rabbit | |
Affinity chromatography | |
RUO | |
24069, 361769, 51684 | |
-20°C | |
Liquid |
Western Blot | |
C.60.7 | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and <0.02% sodium azide; pH 7.5 | |
Q9UMX1, Q9Z0P7 | |
SUFU | |
Synthetic peptide corresponding to residues surrounding Leu458 of human SUFU | |
100 μL | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |