Learn More
SUGT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA528237
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, HeLa. Predicted reactivity: Mouse (86%), Rat (86%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SUGT1 Polyclonal antibody specifically detects SUGT1 in Human samples. It is validated for Immunocytochemistry, Western Blot.Spezifikation
SUGT1 | |
Polyclonal | |
Unconjugated | |
SUGT1 | |
2410174K12Rik; 40-6-3; Aa1114; LOC101122608; MGC137150 protein; Protein 40-6-3; Protein SGT1 homolog; putative 40-6-3 protein; Sgt1; SGT1 homolog, MIS12 kinetochore complex assembly cochaperone; SGT1, suppressor of G2 allele of SKP1; SGT1, suppressor of G2 allele of SKP1 (S. cerevisiae); SGT1B protein; SUGT1; suppressor of G2 allele of SKP1; suppressor of G2 allele of SKP1 homolog; suppressor of G2 allele of SKP1, S. cerevisiae, homolog of; suppressor of g2 allele of SKP1-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10910 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y2Z0 | |
SUGT1 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 295 of Human SUGT1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |