missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF280A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA537067
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 61 kDa. Purity is >95% by SDS-PAGE.
ZNF280A Polyclonal antibody specifically detects ZNF280A in Human, Mouse, Rat samples. It is validated for Western Blot.Spezifikation
ZNF280A | |
Polyclonal | |
Unconjugated | |
ZNF280A | |
3'OY11.1; SUHW1; Suppressor of hairy wing homolog 1; zinc finger protein 280; zinc finger protein 280A; Zinc finger protein 636; ZNF280; ZNF280A; ZNF636 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
129025 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
P59817 | |
ZNF280A | |
Synthetic peptide corresponding to amino acids 491-536 of human SUHW1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |