Learn More
SULT1A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529766
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: HepG2. Predicted reactivity: Rat (81%), Dog (87%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SULT1A1 Polyclonal antibody specifically detects SULT1A1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
SULT1A1 | |
Polyclonal | |
Unconjugated | |
SULT1A1 | |
AI266890; aryl sulfotransferase; Aryl sulfotransferase 1; Aryl sulfotransferase cytosolic 1A phenol-preferring member 3; Aryl sulfotransferase cytosolic, 1A, phenol-preferring, member 3; aryl sulfotransferase IV; arylsulfotransferase ST1A4; ASTIV; CSPS; HAST1/HAST2; MGC131921; MGC5163; Minoxidil sulfotransferase; mSTp1; Mx-ST; OK/SW-cl.88; OTTHUMP00000162568; OTTHUMP00000162569; P PST; P PST 1; Phenol sulfotransferase; Phenol sulfotransferase 1; Phenol sulfotransferase 1a1; phenol/aryl sulfotransferase; phenol-sulfating phenol sulfotransferase 1; P-PST; P-PST 1; PST; PST-1; St1a1; ST1A3; ST1A4; Stm; STP; STP1; sulfokinase; Sulfotransferase 1A1; sulfotransferase family 1A member 1; sulfotransferase family 1A, phenol-preferring, member 1; sulfotransferase family, cytosolic, 1A, phenol preferring; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 1; sulfotransferase, phenol preferring 1; sulphotransferase family cytosolic 1A phenol-prefering member 1; SULT1A1; Sult1a3; The | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6817 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
P50225 | |
SULT1A1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 196 of Human SULT1A1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |