missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SULT1C3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA524548
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
SULT1C3 Polyclonal antibody specifically detects SULT1C3 in Human samples. It is validated for Western Blot.Spezifikation
SULT1C3 | |
Polyclonal | |
Unconjugated | |
SULT1C3 | |
ST1C3; Sulfotransferase 1C3; sulfotransferase family 1C member 3; sulfotransferase family, cytosolic, 1C, member 3; SULT1C3 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
442038 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q6IMI6 | |
SULT1C3 | |
KLH conjugated synthetic peptide between 264-292 amino acids from the C-terminal region of human SULT1C3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |