missing translation for 'onlineSavingsMsg'
Learn More

SULT2A1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA512243

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 10863601

  • 468.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

SULT2A1 Polyclonal antibody specifically detects SULT2A1 in Human, Mouse samples. It is validated for Western Blot.
Spezifikation

Spezifikation

SULT2A1
Polyclonal
Unconjugated
SULT2A1
alcohol sulfotransferase; alcohol/hydroxysteroid sulfotransferase; bile salt sulfotransferase; Bile salt sulfotransferase 1; bile-salt sulfotranasferase 2A1; bile-salt sulfotransferase 2A1; Dehydroepiandrosterone sulfotransferase; DHEAS; DHEA-ST; HST; hSTa; Hydroxysteroid sulfotransferase; hydroxysteroid sulfotransferase a; mSTa1; Smp-2; ST; ST2; ST-20; ST2A1; ST2A3; STa; Sta1; Std; Sth1; Sth2; Sulfotransferase 2A1; sulfotransferase family 2A member 1; sulfotransferase family 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA) -preferring, member 1; sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1; sulfotransferase hydroxysteroid gene 2; sulfotransferase, DHEA preferring; sulfotransferase, hydroxysteroid preferring 1; sulfotransferase, hydroxysteroid preferring 2; SULT2A1
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
20859, 6822
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
2 mg/mL
PBS with 0.09% sodium azide
P52843, Q06520
SULT2A1
KLH conjugated synthetic peptide between 253-285 amino acids from the C-terminal region of human SULT2A
400 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote