missing translation for 'onlineSavingsMsg'
Learn More

SUMF2 Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA547394

Weitere Details : Netto-Gewicht : 0.00010kg

Artikelnummer. 15587882

  • 473.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant mouse SUMF2 is observed and less than 1% cross-reactivity with recombinant human SUMF1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

SUMF2 Polyclonal antibody specifically detects SUMF2 in Human samples. It is validated for Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

SUMF2
Polyclonal
Unconjugated
SUMF2
2610040F05Rik; C-alpha-formyglycine-generating enzyme 2; C-alpha-formylglycine-generating enzyme 2; DKFZp566I1024; DKFZp686I1024; DKFZp686L17160; DKFZp781L1035; FGE2; Inactive C-alpha-formylglycine-generating enzyme 2; MGC99485; Paralog of formylglycine-generating enzyme; paralog of the formylglycine-generating enzyme; pFGE; PSEC0171; sulfatase modifying factor 2; sulfatase-modifying factor 2; Sumf2; UNQ1968/PRO4500
Goat
Antigen affinity chromatography
RUO
25870
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 5% trehalose and No Preservative
Q8NBJ7
SUMF2
Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote