Learn More
SUMO1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA529184
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA5-29184 targets SUMO1 in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-29184 immunogen is recombit fragment corresponding to a region within amino acids 1 and 101 of Human SUMO1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
SUMO1 is an ubiquitin-like protein that can be covalently attached to proteins as a monomer or a lysine-linked polymer. Covalent attachment, via an isopeptide bond, to its substrates requires prior activation by the E1 complex SAE1-SAE2 and linkage to the E2 enzyme UBE2I, and can be promoted by E3 ligases such as PIAS1-4, RANBP2 or CBX4. This post-translational modification on lysine residues of proteins plays a crucial role in a number of cellular processes such as nuclear transport, DNA replication and repair, mitosis and signal transduction. SUMO1 is involved, for instance, in targeting RANGAP1 to the nuclear pore complex protein RANBP2. Polymeric SUMO1 chains are also susceptible to polyubiquitination which functions as a signal for proteasomal degradation of modified proteins. SUMO1 may also regulate a network of genes involved in palate development. Mutations in the gene can result in non-syndromic orofacial cleft 10.Spezifikation
SUMO1 | |
Polyclonal | |
Unconjugated | |
P63165 | |
SUMO1 | |
Recombit fragment corresponding to a region within amino acids 1 and 101 of Human SUMO1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
SUMO1 | |
DAP1, GMP1, OFC10, PIC1, SENP2, SMT3, SMT3C, SMT3H3, UBL1, GAP modifying protein 1, SMT3 homolog 3, sentrin, small ubiquitin-related modifier 1, ubiquitin-homology domain protein PIC1, ubiquitin-like protein SMT3C, ubiquitin-like protein UBL1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7341 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |