missing translation for 'onlineSavingsMsg'
Learn More

SUMO2/SUMO3 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA511319

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 10803641

  • 472.00 € / 100 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

This antibody is predicted to react with bovine, chicken, hamster, porcine, non-human primate, rat and Xenopus and zebrafish based on sequence homology.

SUMO2/SUMO3 Polyclonal antibody specifically detects SUMO2/SUMO3 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

SUMO2/SUMO3
Polyclonal
Unconjugated
Sumo3
2810014B19Rik; D10Ertd345e; GMP1; HSMT3; I79_000932; MIF2 suppressor; Sentrin; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 3; small ubiquitin-like modifier 3 L homeolog; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 3; Smt3; SMT3 (supressor of mif two, 3) homolog 1; SMT3 homolog 1; SMT3 homolog 2; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 suppressor of mif two 3 homolog 3 (yeast); SMT3 supressor of mif two 3 homolog 2; SMT3A; Smt3B; SMT3C; SMT3H1; SMT3H2; SMT3H3; SUMO-1; SUMO2; SUMO-2; Sumo3; SUMO-3; sumo3.L; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; UBL1
Rabbit
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
170930, 20610, 6612, 6613
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P55854, P61956, P61957, Q9Z172
SUMO2, Sumo3
KLH conjugated synthetic peptide between 49-81 amino acids from the C-terminal region of human SUMO2/3
400 μL
Primary
Human, Mouse
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote