Learn More
SUMO2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527636
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: IMR32, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%), Japanese Medaka (97%), Xenopus laevis (98%), Dog (100%), Pig (100%), Chicken (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SUMO2 Polyclonal antibody specifically detects SUMO2 in Human, Mouse samples. It is validated for Western Blot.Spezifikation
SUMO2 | |
Polyclonal | |
Unconjugated | |
SUMO2 | |
HSMT3; sentrin 2; sentrin-2; small ubiquitin-like modifier 2; small ubiquitin-like modifier 2 L homeolog; small ubiquitin-like modifier 3; small ubiquitin-related modifier 2; Small ubiquitin-related modifier 2-A; small ubiquitin-related modifier 3; SMT3 homolog 1; SMT3 homolog 2; SMT3 suppressor of mif two 3 homolog 1; SMT3 suppressor of mif two 3 homolog 2; SMT3 suppressor of mif two 3 homolog 2 (yeast); SMT3 suppressor of mif two 3 homolog 3; SMT3 supressor of mif two 3 homolog 2; Smt3A; Smt3b; SMT3H1; SMT3H2; Sumo2; SUMO-2; sumo2.L; SUMO-2-A; sumo2-a; sumo2-b; SUMO3; SUMO-3; Ubiquitin-like protein SMT3A; ubiquitin-like protein SMT3B; XELAEV_18044053mg | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
170930, 6613 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.56 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P61956, P61957 | |
SUMO2 | |
Recombinant protein encompassing a sequence within the center region of human SUMO2/SUMO3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |