Learn More
SOD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA118278
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
PA1-18278 detects Superoxide Dismutase 3 from human samples. PA1-18278 has been successfully used in immunohistochemistry (paraffin and frozen) and Western blot procedures. The PA1-18278 immunogen is a synthetic peptide corresponding to residues W(18)T G E D S A E P N S D S A E W I R D(37) of human extracellular superoxide dismutase (SOD3), conjugated to diphtheria toxin. Reconstitute with 100 μL of distilled water.
Superoxide dismutase (SOD) is an antioxidant enzyme involved in the defense system against reactive oxygen species (ROS). SOD catalyzes the dismutation reaction of superoxide radical anion (O2) to hydrogen peroxide, which is then catalyzed to innocuous O2 and H2O by glutathione peroxidase and catalase. Several classes of SOD have been identified. These include intracellular copper, zinc SOD (Cu, Zn-SOD/SOD-1), mitochondrial manganese SOD (Mn-SOD/SOD-2) and extracellular Cu, Zn-SOD (EC-SOD/SOD-3) (1). SOD1 is found in all eukaryotic species as a homodimeric 32 kDa enzyme containing one each of Cu and Zn ion per subunit (2). The manganese containing 80 kDa tetrameric enzyme SOD2, is located in the mitochondrial matrix in close proximity to a primary endogenous source of superoxide, the mitochondrial respiratory chain (3). SOD3 is a heparin-binding multimer of disulfide-linked dimers, primarily expressed in human lungs, vessel walls and airways (4). SOD4 is a copper chaperone for superoxide dismutase (CCS), which specifically delivers Cu to copper/zinc superoxide dismutase. CCS may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactorSpezifikation
SOD3 | |
Polyclonal | |
whole serum with no preservative | |
P08294 | |
SOD3 | |
Synthetic peptide corresponding to residues W(18)T G E D S A E P N S D S A E W I R D(37) of human extracellular superoxide dismutase (SOD3). | |
RUO | |
6649 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
SOD3 | |
SOD3, ALS, ALS1, Cu/Zn superoxide dismutase | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |