missing translation for 'onlineSavingsMsg'
Learn More

SUFU Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA529952

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13444347

  • 450.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

PA5-29952 targets Suppressor of Fused in IF, IHC (P), and WB applications and shows reactivity with Human samples. The PA5-29952 immunogen is recombit fragment corresponding to a region within amino acids 267 and 484 of Human Suppressor of Human Fused. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

SUFU encodes a component of the Sonic hedgehog (SHH; MIM 600725)/Patched (PTCH; MIM 601309) signaling pathway. Mutations in genes encoding components of this pathway are deleterious for normal development and are associated with cancer-predisposing syndromes (e.g., HPE3, MIM 142945; BCNS, MIM 109400; and GCPS, MIM 175700).
Spezifikation

Spezifikation

SUFU
Polyclonal
Unconjugated
Q9UMX1, Q9Z0P7
SUFU
Recombit fragment corresponding to a region within amino acids 267 and 484 of Human Suppressor of Human Fused
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
SUFU
PRO1280, SUFUH, SUFUXL, suppressor of fused homolog
Rabbit
Antigen affinity chromatography
RUO
51684, 24069
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote