missing translation for 'onlineSavingsMsg'
Learn More

SUPV3L1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marque:  Invitrogen PA532082

Informations supplémentaires : Netto-Gewicht : 0.01000kg

Code produit. 13406657

  • 471.00 € / 100 Mikroliter
Verfügbar ab: 22-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Description

Description

Recommended positive controls: PC-3, HeLa. Predicted reactivity: Mouse (83%), Rat (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SUPV3L1 Polyclonal antibody specifically detects SUPV3L1 in Human samples. It is validated for Immunocytochemistry, Western Blot.
Spécification

Spécification

SUPV3L1
Polyclonal
Unconjugated
SUPV3L1
6330443E10Rik; ATP-dependent RNA helicase SUPV3L1, mitochondrial; Suppressor of var1 3-like protein 1; suppressor of var1, 3-like 1; suppressor of var1, 3-like 1 (S. cerevisiae); suppressor of var1, 3-like 1(SUV3); SUPV3L1; SUV3; Suv3 like RNA helicase; Suv3l1; SUV3-like helicase; SUV3-like protein 1
Rabbit
Antigen affinity chromatography
RUO
6832
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Western Blot
1 mg/mL
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Q8IYB8
SUPV3L1
Recombinant fragment corresponding to a region within amino acids 545 and 779 of Human SUPV3L1
100 μL
Primary
Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions