missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SURF1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA536198
Weitere Details : Netto-Gewicht : 0.00910kg
Beschreibung
This antibody detects endogenous protein at a molecular weight of 33 kDa. Purity is >95% by SDS-PAGE.
SURF1 Polyclonal antibody specifically detects SURF1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
SURF1 | |
Polyclonal | |
Unconjugated | |
SURF1 | |
0610010F23Rik; Ab1-205; CMT4K; Surf1; Surf-1; surfeit 1; surfeit gene 1; surfeit locus protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20930, 64463, 6834 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P09925, Q15526, Q9QXU2 | |
SURF1 | |
Synthetic peptide corresponding to amino acids 150-200 of human SURF-1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |