missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SFTPA1/SFTPA2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA538059
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
This antibody is predicted to react with cow based on sequence homology.
SFTPA1/SFTPA2 Polyclonal antibody specifically detects SFTPA1/SFTPA2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
SFTPA1/SFTPA2 | |
Polyclonal | |
Unconjugated | |
SFTPA2 | |
35 kDa pulmonary surfactant-associated protein; Alveolar proteinosis protein; COLEC4; COLEC5; collectin 5; collectin-4; Collectin-5; FLJ50593; FLJ51913; FLJ61144; FLJ77898; FLJ79095; FLJ99559; MGC133365; MGC198590; PSAP; PSPA; PSP-A; pulmonary surfactant-associated protein A; Pulmonary surfactant-associated protein A1; pulmonary surfactant-associated protein A2; Sftp1; Sftp-1; Sftpa; Sftpa1; SFTPA1B; SFTPA2; SFTPA2B; Sftpl; SP-2A; SP-2A beta; SP-2A gamma; SPA; SP-A; SPA1; SP-A1; SP-A1 beta; SP-A1 delta; SP-A1 epsilon; SP-A1 gamma; SPA2; SP-A2; SP-A2 alpha; SP-A2 delta; SPAII; surfactant associated protein A; surfactant associated protein A1; surfactant protein A1; surfactant protein A1 variant AB'D' 6A; surfactant protein A1 variant AB'D' 6A2; surfactant protein A1 variant AB'D' 6A3; surfactant protein A1 variant AB'D' 6A4; surfactant protein A1 variant ACD' 6A; surfactant protein A1 variant ACD' 6A2; surfactant protein A1 variant ACD' 6A3; surfactant protein A1 variant ACD' 6A4; surfa | |
Goat | |
Antigen affinity chromatography | |
RUO | |
653509, 729238 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8IWL1, Q8IWL2 | |
Sftpa1, SFTPA2 | |
Peptide with sequence C-HLDEELQATLHDFR, from the internal region of SFTPA1 (aa102-115) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |