missing translation for 'onlineSavingsMsg'
Learn More

SUSD2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA533064

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 13404739

  • 473.00 € / 50 Mikrogramm
Verfügbar ab: 23-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey (100%); Chimpanzee, Orangutan (94%); Gibbon, Panda (89%); Galago, Opossum (83%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.

SUSD2 Polyclonal antibody specifically detects SUSD2 in Human, Non-human Primate samples. It is validated for Immunohistochemistry (Paraffin).
Spezifikation

Spezifikation

SUSD2
Polyclonal
Unconjugated
SUSD2
BK65A6.2; SUSD2; Sushi domain (SCR repeat) containing; sushi domain containing 2; sushi domain-containing protein 2; testicular tissue protein Li 190
Rabbit
Antigen affinity chromatography
RUO
56241
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q9UGT4
SUSD2
Synthetic 18 amino acid peptide from internal region of human SUSD2.
50 μg
Primary
Human, Non-human Primate
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote