missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SUSD3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544351
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: LRLPPQATFQ VLRGNGASVG TVLMFRCPSN HQMVGSGLLT CTWKGSIAEW Sequence homology: Cow: 77%; Human: 100%; Pig: 77%
SUSD3 Polyclonal antibody specifically detects SUSD3 in Human samples. It is validated for Western Blot.Spezifikation
SUSD3 | |
Polyclonal | |
Unconjugated | |
SUSD3 | |
1700017I11Rik; 2810440J20Rik; Susd3; sushi domain containing 3; sushi domain-containing protein 3; UNQ9387/PRO34275 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
203328 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96L08 | |
SUSD3 | |
Synthetic peptide directed towards the N-terminal of human SUSD3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |