Learn More
SUZ12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen 491035
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
This zinc finger gene has been identified at the breakpoints of a recurrent chromosomal translocation reported in endometrial stromal sarcoma. Recombination of these breakpoints results in the fusion of this gene and JAZF1. The protein encoded by this gene contains a zinc finger domain in the C terminus of the coding region.
SUZ12 Polyclonal antibody specifically detects SUZ12 in Human, Mouse samples. It is validated for ChIP Assay, Western Blot.Spezifikation
SUZ12 | |
Polyclonal | |
Unconjugated | |
SUZ12 | |
2610028O16Rik; AI195385; AU016842; AW536442; chET 9 protein; CHET9; chromatin precipitated E2F target 9 protein; D11Ertd530e; JJAZ1; Joined to JAZF1 protein; Kiaa0160; mKIAA0160; Polycomb protein Suz12; suppressor of zeste 12 homolog; suppressor of zeste 12 homolog (Drosophila); suppressor of zeste 12 protein homolog; Suz12; SUZ12 polycomb repressive complex 2 subunit | |
Rabbit | |
Protein G | |
RUO | |
23512 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ChIP Assay, Western Blot | |
2 mg/mL | |
PBS with 0.05% ProClin 300, 0.05% sodium azide | |
Q15022, Q80U70 | |
SUZ12 | |
Raised against the N-terminus of the recombinant human Suz12 protein. | |
50 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |