missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SV40 T-Antigen Monoclonal Antibody (PAb108), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA512463
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody reacts with the large T and small T-antigen of SV40 under native and denatured conditions. It will also react with the large T-antigen of BKV, human papovavirus.
SV40 T-Antigen Monoclonal antibody specifically detects SV40 T-Antigen in Virus samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Spezifikation
SV40 T-Antigen | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
Virus | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
PAb108 | |
Unconjugated | |
simian vacuolating virus 40; simian virus; Simian virus 40; SV40 T-antigen; sv40 virus large t antigen | |
SV40 transformed mouse line B4 | |
500 μL | |
Primary | |
4° C | |
Liquid |