missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SWAP70 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA141490
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Suggested positive control: Daudi.
SWAP70 Polyclonal antibody specifically detects SWAP70 in Human samples. It is validated for Western Blot.Spezifikation
SWAP70 | |
Polyclonal | |
Unconjugated | |
SWAP70 | |
70kDa; AV235546; FLJ39540; HSPC321; Kiaa0640; KIAA0640 protein; SWA-70 protein; SWAP complex protein; SWAP complex protein, 70 kDa; SWAP switching B-cell complex 70; SWAP switching B-cell complex 70kDa subunit; SWAP70; SWAP-70; SWAP-70 protein; Switch-associated protein 70 | |
Rabbit | |
Protein G | |
RUO | |
23075 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.2% gelatin and 0.05% sodium azide | |
Q9UH65 | |
SWAP70 | |
Synthetic peptide corresponding to residues 300-350 of human SWAP-70. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |