missing translation for 'onlineSavingsMsg'
Learn More

Syk Polyclonal Antibody, Invitrogen™

Goat Polyclonal Antibody

Marke:  Invitrogen PA186631

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11513791

  • 548.00 € / 100 Mikrogramm
Verfügbar ab: 08-05-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Predicted to react with dog, mouse and rat based on sequence homology. In Western blot, this antibody detects a band at ∽70 kDa in human bone marrow, lymph node and spleen lysates. Goat anti Syk antibody detects an epitope within the internal region of SYK, a approximately 70 kDa non-receptor protein tyrosine kinase whose role differs in haematopoietic and non haematopoietic stem cells.

Syk Polyclonal antibody specifically detects Syk in Human samples. It is validated for ELISA, Western Blot.
Spezifikation

Spezifikation

Syk
Polyclonal
Unconjugated
Syk
DKFZp313N1010; EC 2.7.10.2; FLJ25043; FLJ37489; HGNC:11205; kinase Syk; KSYK; p72Syk; p72-Syk; ptk72; SP1; spleen associated tyrosine kinase; spleen tyrosine kinase; SYK; syk signaling; Sykb; tyrosine-kinase syk; tyrosine-protein kinase SYK
Goat
Affinity chromatography
RUO
6850
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P43405
Syk
peptide with sequence C-KYLQQNRHVKDKN from the internal region of the SYK sequence
100 μg
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote