missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYNGR4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518327
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
SYNGR4 Polyclonal antibody specifically detects SYNGR4 in Human samples. It is validated for Western Blot.Spezifikation
SYNGR4 | |
Polyclonal | |
Unconjugated | |
Syngr4 | |
1700016O14Rik; MGC125805; synaptogyrin 4; synaptogyrin 4 protein; synaptogyrin-4; SYNGR4; testis tissue sperm-binding protein Li 72n | |
Goat | |
Antigen affinity chromatography | |
RUO | |
23546 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O95473 | |
Syngr4 | |
Synthetic peptide sequence (HIPKSLQELANS) corresponding to the N-terminus amino acids of SYNGR4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |