Learn More
Synaptojanin 1 Monoclonal Antibody (AC1), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA3936
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA3-936 detects Synaptojanin 1 from rat and human samples. MA3-936 has been successfully used in Western blotting, IHC (frozen), IP and ICC/IF procedures. By Western blot MA3-936 detects a band at 170 kDa representing Synaptojanin 1. The MA3-936 immunogen is proline rich domain of a fusion protein comprising aa 1156-1286 of rat synaptojanin 1.
Synaptojanin 1 Monoclonal antibody specifically detects Synaptojanin 1 in Human, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
Synaptojanin 1 | |
Monoclonal | |
Conc. Not Determined | |
ascites diluted in PBS with 0.05% sodium azide | |
O43426, Q62910 | |
SYNJ1 | |
Proline rich domain of a fusion protein comprising amino acids 1156-1286 of rat synaptojanin 1. | |
RUO | |
85238, 8867 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
AC1 | |
Unconjugated | |
SYNJ1 | |
A930006D20Rik; AA675315; inositol 5'-phosphatase (synaptojanin 1); inositol polyphosphate-5-phosphatase G; INPP5G; KIAA0910; mKIAA0910; PARK20; phosphoinositide 5-phosphatase; synaptic inositol 1,4,5-trisphosphate 5-phosphatase 1; synaptic inositol-1; synaptic inositol-1,4,5-trisphosphate 5-phosphatase 1; synaptojanin 1; synaptojanin-1; synaptojanin-1, polyphosphoinositide phosphatase; Synj1 | |
Mouse | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG1 |