Learn More
Synaptophysin Monoclonal Antibody (SP11), Invitrogen™
Rabbit Monoclonal Antibody
Marke: Invitrogen MA139558
Weitere Details : Netto-Gewicht : 0.10000kg
Beschreibung
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is pancreas or neuroendocrine tumor.
Synaptophysin Monoclonal antibody specifically detects Synaptophysin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Synaptophysin | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant diluted in TBS with 1% BSA and 0.1% sodium azide; pH 7.5 | |
P07825, P08247, Q62277 | |
Syp | |
Synthetic peptide corresponding to C-terminus of human synaptophysin. | |
RUO | |
20977, 24804, 6855 | |
-20°C or -80°C if preferred | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
SP11 | |
Unconjugated | |
Syp | |
A230093K24Rik; AI848995; BM89 antigen; I79_012252; Major synaptic vesicle protein p38; MRX96; MRXSYP; p38; Syn; Synaptophysin; Syp; Syp I; Syp1 | |
Rabbit | |
1 mL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |