missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX16 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSS00014W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water.
Spezifikation
STX16 | |
Polyclonal | |
Stx16 | |
Syntaxin 16, STX16, Syn16 | |
Rabbit | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry | |
Unconjugated | |
Q8BVI5 | |
STX16 | |
A synthetic peptide from mouse Syntaxin-16 | |
RUO | |
228960 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |