missing translation for 'onlineSavingsMsg'
Learn More

STX16 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Invitrogen PA519592

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11397543

Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen


Beschreibung

Beschreibung

Not suitable for use with Immunocytochemistry. Not suitable for use with Immunofluorescence.

This gene encodes a protein that is a member of the syntaxin or t-SNARE family. These proteins are found on cell membranes and serve as the targets for V-SNARES permitting specific synaptic vesicle docking and fusion. A microdeletion in the region of chromosome 20 where this gene is located has been associated with pseudohypoparathyroidism type Ib. Multiple transcript variants have been found for this gene. Read-through transcription also exists between this gene and the neighboring downstream aminopeptidase-like 1 gene.
Spezifikation

Spezifikation

STX16
Polyclonal
Unconjugated
RP11-261P9.3, MGC90328, SYN16, syntaxin-16
Rabbit
Antigen affinity chromatography
RUO
33034
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
Syx16
Syx16
Synthetic peptide conjugated to KLH derived from within residues 200 - 300 of Fruit fly (Drosophila melanogaster) Syntaxin 16.
100 μg
Primary
Drosophila
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote