missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX1A/STX1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSS00013W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water.
The protein encoded by this gene is a synaptotagmin-like protein (SLP) that belongs to a C2 domain-containing protein family. The SLP homology domain (SHD) of this protein has been shown to specifically bind the GTP-bound form of Ras-related protein Rab-27A (RAB27A). This protein plays a role in RAB27A-dependent vesicle trafficking and controls melanosome distribution in the cell periphery. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Spezifikation
STX1A/STX1B | |
Polyclonal | |
STX1A | |
STX1A, HGNC:11433, syntaxin 1A, HPC-1, STX1, p35-1, adipocyte, C1Q and collagen domain containing, adiponectin, adipose most abundant gene transcript 1, syntaxin 1B2, STX1B, syntaxin 1B, STX1B1, STX1B2, FP17469, syntaxin 1B1, syntaxin a/b | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
P61266, Q16623, O35526, P61264, P32851, P61265 | |
STX1A, STX1B | |
A synthetic peptide from mouse Syntaxin-1A conjugated to an immunogenic carrier protein | |
RUO | |
112755, 116470, 20907, 24923, 56216, 6804 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |