Learn More
Syntaxin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA544996
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: VEMQGEMINR IEKNILSSAD YVERGQEHVK TALENQKKAR KKKVLIAICV Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 91%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%
Syntaxin 4 Polyclonal antibody specifically detects Syntaxin 4 in Human samples. It is validated for Western Blot.Spezifikation
Syntaxin 4 | |
Polyclonal | |
Unconjugated | |
STX4 | |
hypothetical protein LOC508675; p35-2; Renal carcinoma antigen NY-REN-31; Stx4; STX4A; Syn4; Syn-4; syntaxin 4; syntaxin 4A (placental); syntaxin-4; syntaxin-4 isoform A | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6810 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q12846 | |
STX4 | |
Synthetic peptide directed towards the C-terminal of human STX4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |