missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STX7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen OSS00012W
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Reconstitute with 100 μL of distilled water.
Spezifikation
STX7 | |
Polyclonal | |
Stx7 | |
Syntaxin-7, STX7, Syn7 | |
Rabbit | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
O70439, O70257 | |
STX7 | |
A synthetic peptide from n-terminal region of mouse Syntaxin-7 | |
RUO | |
53331, 60466 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |