missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntenin 1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518595
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with canine, mouse and rat based on sequence homology.
Syntenin 1 Polyclonal antibody specifically detects Syntenin 1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
Syntenin 1 | |
Polyclonal | |
Unconjugated | |
SDCBP | |
MDA9; MDA-9; melanoma differentiation associated protein-9; melanoma differentiation-associated protein 9; Pro-TGF-alpha cytoplasmic domain-interacting protein 18; sb:eu862; Scaffold protein Pbp1; sdcbp; si:dkey-235k4.1; ST1; Sycl; syndecan binding protein; syndecan binding protein (syntenin); syndecan interacting protein; syndecan-binding protein; syndecan-binding protein 1; syntenin; syntenin 1; syntenin-1; syntenin-b; TACIP18; Unknown (protein for MGC:128349) | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6386 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O00560 | |
SDCBP | |
Synthetic peptide sequence (SLEDLKVDKVIQAQ) corresponding to the N-terminus amino acids of SDCBP | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |