missing translation for 'onlineSavingsMsg'
Learn More

SNTB2 Monoclonal Antibody (1351), Invitrogen™

Mouse Monoclonal Antibody

Marke:  Invitrogen MA1745

Weitere Details : Netto-Gewicht : 0.01000kg

Artikelnummer. 11590602

  • 449.00 € / 100 Mikrogramm
Verfügbar ab: 19-04-2024
Sie sich ein, um den Lagerbestand zu sehen

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

MA1-745 detects alpha 1, beta 1 and beta 2 subunits of syntrophin. This antibody has been shown to react with human, mouse, rat, chicken, canine and amphibian samples. MA1-745 has been successfully used in Western blot, immunoprecipitation and immunofluorescence procedures. By Western blot, this antibody detects an ∽58 kDa protein representing syntrophin from rat and mouse cell extracts. The MA1-745 immunogen is whole purified syntrophin from Torpedo californica electric organ postsynaptic membrane. Characterization studies of this antibody have demonstrated that clone 1351 is directed against an epitope within the PDZ domain of syntrophin. MA1-745, clone 1351, is seen as the "e;gold standard"e; for syntrophin assessment.

SNTB2 Monoclonal antibody specifically detects SNTB2 in Amphibian, Canine, Chicken, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot.
Spezifikation

Spezifikation

SNTB2
Monoclonal
1 mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide
Q13425, Q61235
SNTB2
Whole purified syntrophin from Torpedo californica electric organ postsynaptic membrane.
100 μg
Primary
Amphibian, Canine, Chicken, Human, Mouse, Rat
Antibody
IgG1
Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot
1351
Unconjugated
SNTB2
59 kDa dystrophin-associated protein A1 basic component 2; beta-2-syntrophin; D16S2531E; dystrophin-associated protein A1, 59kD, basic component 2; EST25263; Snt2; SNT2B2; SNT3; SNTB2; SNTL; SYN; syntrophin beta 2; syntrophin, basic 2; syntrophin, beta 2; syntrophin, beta 2 (dystrophin-associated protein A1, 59kDa, basic component 2); syntrophin-3; Syntrophin-like
Mouse
Protein G
RUO
20650, 479675, 6645, 689421, 769654
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote