missing translation for 'onlineSavingsMsg'
Learn More
Learn More
T-bet Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA519773
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
For Western Blot, this antibody has non-specific bands at 70 kDa.
T-bet Polyclonal antibody specifically detects T-bet in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot.Spezifikation
T-bet | |
Polyclonal | |
Unconjugated | |
Tbx21 | |
HGNC:11599; LOW QUALITY PROTEIN: T-box transcription factor TBX21; Tbet; T-bet; TBLYM; T-box 21; T-box expressed in T cells; T-box protein 21; T-box transcription factor 21; T-box transcription factor TBX21; TBT1; TBX21; T-cell-specific T-box transcription factor T-bet; T-PET; transcription factor TBLYM; transcription factor T-box 21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30009, 57765 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunoprecipitation, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.02% sodium azide; pH 7.4 | |
Q9JKD8, Q9UL17 | |
Tbx21 | |
Synthetic peptide conjugated to KLH derived from within residues 200 - 300 of Human T-bet / Tbx21. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |