Learn More
CTNNA3 Monoclonal Antibody (892_24D2S), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA106311
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
MA1-06311 detects alphaT-catenin in human samples. MA1-06311 has sucessfully been used in Western blotting, immunoprecipitation, immunohistochemistry, and immunocytochemistry. By Western blotting, it detects a ∽300 kDa protein representing alphaT-catenin. The MA1-06311 immunogen is a synthetic peptide corresponding to residues C V(164) A N K S D L Q K T Y Q K L(177) of the human T-catenin protein with N-term Cys, coupled to KLH.
CTNNA3 Monoclonal antibody specifically detects CTNNA3 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
CTNNA3 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q9UI47 | |
CTNNA3 | |
Synthetic peptide to corresponding residues C V(164) A N K S D L Q K T Y Q K L(177) of the human T-catenin protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
892_24D2S | |
Unconjugated | |
CTNNA3 | |
4930429L08Rik; 4933408A16; Alpha T-catenin; Alpha-3 catenin; alpha-catenin-like protein; alpha-T-catenin; ARVD13; Cadherin-associated protein; catenin (cadherin associated protein), alpha 3; catenin (cadherin-associated protein), alpha 3; catenin alpha 3; catenin alpha-3; Catna3; CTNNA3; RGD1559455; RGD1562230; Vr22 | |
Mouse | |
purified | |
RUO | |
29119 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |