missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS1R3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA534256
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla (100%) Orangutan (90%) Marmoset (85%) Baboon, Monkey, Panda (80%).
TAS1R3 Polyclonal antibody specifically detects TAS1R3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
TAS1R3 | |
Polyclonal | |
Unconjugated | |
TAS1R3 | |
Sac; Saccharin preference protein; sweet taste receptor T1R3; T1r3; TAS1R3; taste 1 receptor member 3; taste receptor type 1 member 3; taste receptor, type 1, member 3; TR3; TS1R3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83756 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q7RTX0 | |
TAS1R3 | |
Synthetic 20 amino acid peptide from N-terminal extracellular domain of human TAS1R3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |