Learn More
T7 Tag Goat anti-Tag, FITC, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Marke:  Invitrogen PA132390
Weitere Details : Netto-Gewicht : 0.01000kg
Artikelnummer. 11562412
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Beschreibung
Beschreibung
PA1-32390 detects T7 from rat samples. PA1-32390 has been successfully used in immunocytochemistry, immunohistochemistry applications. The PA1-32390 immunogen is: Goats were immunized with T7 cleavage site (MASMTGGQQMG). Antibody was isolated by affinity chromatography using the peptide immobilized on solid support and conjugated to fluorescein isothiocynate (FITC). F/P ration is 3 to 7.
The T7 Tag is an 11 amino acid peptide derived from the bacteriophage T7 major capsid protein, gene 10. T7 cleavage site: MASMTGGQQMG. Epitope tags provide a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available.
Spezifikation
Spezifikation
T7 Tag
Polyclonal
FITC
Goat
Purified
RUO
4° C, store in dark, DO NOT FREEZE!
Liquid
Tag
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
1 mg/mL
MASMTGGQQMG; T7 cleavage
Goats were immunized with T7 cleavage site (MASMTGGQQMG). Antibody was isolated by affinity chromatography using the peptide immobilized on solid support and conjugated to fluorescein isothiocynate (FITC). F/P ration is 3 to 7.