Learn More
TAAR8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA533069
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Galago (83%). For IHC(P), use proteinase K antigen retrieval. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
TAAR8 Polyclonal antibody specifically detects TAAR8 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin).Spezifikation
TAAR8 | |
Polyclonal | |
Unconjugated | |
TAAR8 | |
G protein-coupled receptor 102; Gm1149; Gm230; GPR102; G-protein coupled receptor 102; mTaar8a; Q923; Ta10; Ta11; TA5; Ta7; TAAR8; Taar8a; Taar8b; Taar8c; Tar11; TaR-11; TAR5; TaR-5; TaR-8; TaR-8a; trace amine associated receptor 8; trace amine receptor 11; Trace amine receptor 5; trace amine receptor 8; trace amine receptor 8a; trace amine-associated receptor 8; Trace amine-associated receptor 8a; trace-amine-associated receptor 8a; Trar11; Trar5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83551 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q969N4 | |
TAAR8 | |
Synthetic 12 amino acid peptide from C-Terminus of human TAAR8. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |