Learn More
TAF15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA527845
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji, NIH-3T3. Predicted reactivity: Mouse (94%), Pig (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TAF15 Polyclonal antibody specifically detects TAF15 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Spezifikation
TAF15 | |
Polyclonal | |
Unconjugated | |
TAF15 | |
2610111C21Rik; 68 kDa TATA-binding protein-associated factor; 68kDa; hTAFII68; LOC100713096; Npl3; RBP56; RBP56/CSMF fusion; RNA-binding protein 56; TAF(II)68; Taf15; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68 kDa; TAF15 RNA polymerase II, TATA box binding protein (TBP)-associated factor, 68kDa; TAF2N; TAFII68; TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA binding protein 56); TATA box binding protein (TBP)-associated factor, RNA polymerase II, N, 68kD (RNA-binding protein 56); TATA box binding protein associated factor 15; TATA box-binding protein-associated factor 2N (RNA-binding protein 56); TATA-bin; TATA-binding protein-associated factor 2N; TATA-box binding protein associated factor 15; TBP-associated factor 15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
70439, 8148 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q92804 | |
TAF15 | |
Recombinant fragment corresponding to a region within amino acids 15 and 279 of Human TAFII68 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |