missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAGLN Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marke: Invitrogen PA518252
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
This antibody is predicted to react with bovine and canine based on sequence homology.
TAGLN Polyclonal antibody specifically detects TAGLN in Human samples. It is validated for Western Blot.Spezifikation
TAGLN | |
Polyclonal | |
Unconjugated | |
TAGLN | |
22 kDa actin-binding protein; 25 kDa F-actin-binding protein; actin-associated protein p27; DKFZp686B01212; DKFZp686P11128; Protein WS3-10; similar to transgelin; Sm22; sm22 alpha; SM22A; Sm22alpha; SM22-alpha; SMCC; smooth muscle 22 protein; smooth muscle protein 22-alpha; TAGLN; TAGLN1; Transgelin; transgelin variant 2; Ws310; WS3-10 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
6876 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q01995 | |
TAGLN | |
Synthetic peptide sequence (MTGYGRPRQIIS) corresponding to the C-terminus amino acids of TAGLN | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |