Learn More
TAL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA546840
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Peptide sequence: QDVLSPNSSC GSSLDGAASP DSYTEEPAPK HTARSLHPAM LPAADGAGPR Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 92%
TAL1 Polyclonal antibody specifically detects TAL1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TAL1 | |
Polyclonal | |
Unconjugated | |
TAL1 | |
basic helix-loop-helix transcription factor; bHLHa17; Class A basic helix-loop-helix protein 17; HGNC:11556; LOW QUALITY PROTEIN: T-cell acute lymphocytic leukemia protein 1; Scl; scl/tal1; SCL/tal-1; SCL-alpha; SCL-beta; si:ch211-135j1.3; stem cell leukemia protein; stem cell leukemia protein SCL; stem cell protein; stem-cell leukemia hematopoietic transcription factor; T cell acute lymphocytic leukemia 1; TAL bHLH transcription factor 1, erythroid differentiation factor; TAL1; Tal-1; tal-1 product; T-cell acute lymphocytic leukemia 1; T-cell acute lymphocytic leukemia protein 1; T-cell acute lymphocytic leukemia protein 1 homolog; T-cell leukemia/lymphoma protein 5; TCL5; transcription factor; zgc:85605; zSCL | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6886 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P17542 | |
TAL1 | |
Synthetic peptide directed towards the C-terminal of human TAL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |