missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TANK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA525754
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TANK Polyclonal antibody specifically detects TANK in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TANK | |
Polyclonal | |
Unconjugated | |
Tank | |
C86182; E430026L09Rik; ITRAF; I-TRAF; LOW QUALITY PROTEIN: TRAF family member-associated NF-kappa-B activator; OTTHUMP00000200694; OTTHUMP00000200695; OTTHUMP00000200696; OTTHUMP00000200698; OTTHUMP00000200700; OTTHUMP00000200703; OTTHUMP00000200704; TANK; TANK-like protein; TRAF family member associated NFKB activator; TRAF family member-associated Nf-kappa B activator; TRAF family member-associated NF-kappa-B activator; TRAF family member-associated NFKB activator; TRAF interacting protein TANK; TRAF2; TRAF-interacting protein; zgc:153048 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10010 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q92844 | |
Tank | |
KLH conjugated synthetic peptide between 88-114 amino acids from the central region of human TANK | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |